3-Methyl-3-(1H-pyrrol-1-yl)butanoic acid 95+%

Supplier: Apollo Scientific
Warning

OR7861-1G OR7861-5G OR7861-25G
APOSOR7861-1GEA 185 EUR
APOSOR7861-1G APOSOR7861-5G APOSOR7861-25G
3-Methyl-3-(1H-pyrrol-1-yl)butanoic acid 95+%
3-Methyl-3-(1H-pyrrol-1-yl)butanoic acid
Formula: C₉H₁₃NO₂
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00232890
CAS Number: 395090-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR