4-Bromo-1-methyl-2-(methylsulphonyl)benzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-(methylsulphonyl)toluene
OR7959-1G OR7959-5G
APOSOR7959-1GEA 81.2 EUR
APOSOR7959-1G APOSOR7959-5G
4-Bromo-1-methyl-2-(methylsulphonyl)benzene
4-Bromo-1-methyl-2-(methylsulphonyl)benzene
Formula: C₈H₉BrO₂S
MW: 249.13 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD04037925
CAS Number: 254887-17-7