2,5-Dichlorothiophenol 98%
Supplier: Apollo Scientific
Warning
OR7999-100G OR7999-25G OR7999-5G
APOSOR7999-100GEA 1330 EUR
APOSOR7999-100G APOSOR7999-25G APOSOR7999-5G
2,5-Dichlorothiophenol 98%
2,5-Dichlorothiophenol
Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 112…116 °C/50mmHg
Melting Pt: 28
Density: 1.415
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004831
CAS Number: 5858-18-4
EINECS: 227-486-9