5-Iodo-2-furaldehyde 95%
Supplier: Apollo Scientific
Warning
OR8105-100G OR8105-25G OR8105-5G OR8105-1G
APOSOR8105-100GEA 758 EUR
APOSOR8105-100G APOSOR8105-25G APOSOR8105-5G APOSOR8105-1G
5-Iodo-2-furaldehyde 95%
5-Iodo-2-furaldehyde
Formula: C₅H₃IO₂
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00159503
CAS Number: 2689-65-8