1-(4-Aminophenyl)-4-methylpiperazine 99%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline
OR8114-1G OR8114-25G OR8114-5G
APOSOR8114-1GEA 21.9 EUR
APOSOR8114-1G APOSOR8114-25G APOSOR8114-5G
1-(4-Aminophenyl)-4-methylpiperazine 99%
1-(4-Aminophenyl)-4-methylpiperazine
Formula: C₁₁H₁₇N₃
MW: 191.28 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00172703
CAS Number: 16153-81-4
UN: 3263
ADR: 8 III