2,5-Dibromothiazole 97%
Supplier: Apollo Scientific
Warning
OR8408-1G OR8408-5G OR8408-25G
APOSOR8408-1GEA 0 EUR
APOSOR8408-1G APOSOR8408-5G APOSOR8408-25G
2,5-Dibromothiazole 97%
2,5-Dibromothiazole
Formula: C₃HBr₂NS
MW: 242.92 g/mol
Melting Pt: 45…49 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00016891
CAS Number: 4175-78-4