5-(Morpholinomethyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Dihydro-5-[(morpholin-4-yl)methyl]-2-thioxopyrimidin-4(1H)-one
OR8521-10G OR8521-1G OR8521-5G
APOSOR8521-10GEA 546 EUR
APOSOR8521-10G APOSOR8521-1G APOSOR8521-5G
5-(Morpholinomethyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
5-(Morpholinomethyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
Formula: C₉H₁₃N₃O₂S
MW: 227.29 g/mol
Melting Pt: 223…224 °C
Storage Temperature: Ambient
MDL Number: MFCD01689081
CAS Number: 89665-74-7