9-Bromophenanthrene 98%
Supplier: Apollo Scientific
|
Warning
|
OR8571-25G
OR8571-100G
OR8571-1G
APOSOR8571-25GEA
24.8
EUR
APOSOR8571-25G
APOSOR8571-100G
APOSOR8571-1G
9-Bromophenanthrene 98%
9-Bromophenanthrene
Formula:
C₁₄H₉Br Boiling Pt: 180…190 °C/2mmHg Melting Pt: 60…64 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001174 CAS Number: 573-17-1 EINECS: 209-351-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...