9-Bromoanthracene 98%
Supplier: Apollo Scientific
Warning
OR8575-100G OR8575-25G OR8575-5G
APOSOR8575-100GEA 141 EUR
APOSOR8575-100G APOSOR8575-25G APOSOR8575-5G
9-Bromoanthracene 98%
9-Bromoanthracene
Formula: C₁₄H₉Br
Melting Pt: 97…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3