5-(Hydroxymethyl)-2-iodophenol 97%
Supplier: Apollo Scientific
Warning
OR8637-1G OR8637-10G OR8637-5G OR8637-25G
APOSOR8637-1GEA 66.7 EUR
APOSOR8637-1G APOSOR8637-10G APOSOR8637-5G APOSOR8637-25G
5-(Hydroxymethyl)-2-iodophenol 97%
5-(Hydroxymethyl)-2-iodophenol
Formula: C₇H₇IO₂
MW: 250.04 g/mol
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD06202863
CAS Number: 773869-57-1