Furan-3-boronic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Furanboronic acid, 3-Furanylboronic acid
OR8670-1G OR8670-5G
APOSOR8670-1GEA 21.9 EUR
APOSOR8670-1G APOSOR8670-5G
Furan-3-boronic acid 98%
Furan-3-boronic acid
Formula: C₄H₅BO₃
MW: 111.89 g/mol
Melting Pt: 139…144 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01319007
CAS Number: 55552-70-0