2-Picolyl chloride hydrochloride 98%
Supplier: Apollo Scientific
Danger
OR8686-25G OR8686-5G OR8686-100G OR8686-500G
APOSOR8686-25GEA 21.9 EUR
APOSOR8686-25G APOSOR8686-5G APOSOR8686-100G APOSOR8686-500G
2-Picolyl chloride hydrochloride 98%
2-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
Melting Pt: 119…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00012811
CAS Number: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III