[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
Supplier: Apollo Scientific
Warning
Synonyms: [1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd(dppf)Cl2, 1,1’-Bis(diphenylphosphino)ferrocene palladium(II)dichloride
OR8775-1G OR8775-5G OR8775-25G
APOSOR8775-1GEA 33.5 EUR
APOSOR8775-1G APOSOR8775-5G APOSOR8775-25G
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
[1,1'-Bis(diphenylphosphino)ferrocene]palladium (II) chloride
Catalyst for Suzuki and Stille couplings.
Formula: C₃₄H₂₈Cl₂P₂FePd
MW: 731.70 g/mol
Melting Pt: 266…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00015757
CAS Number: 72287-26-4
EINECS: 460-040-3