β-Chloropropiophenone 90%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloropropiophenone
OR8787-100G OR8787-1KG OR8787-250G OR8787-500G
APOSOR8787-100GEA 26.1 EUR
APOSOR8787-100G APOSOR8787-1KG APOSOR8787-250G APOSOR8787-500G
β-Chloropropiophenone 90%
β-Chloropropiophenone
Formula: (C₆H₅)C(O)CH₂CH₂Cl
MW: 168.62 g/mol
Boiling Pt: 113…115 °C/4mmHg
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000990
CAS Number: 936-59-4
EINECS: 213-317-6