4-(Diethylamino)benzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Diethylaminobenzaldehyde
OR8807-5G OR8807-10G OR8807-25G OR8807-100G
APOSOR8807-5GEA 58.2 EUR
APOSOR8807-5G APOSOR8807-10G APOSOR8807-25G APOSOR8807-100G
4-(Diethylamino)benzaldehyde
4-(Diethylamino)benzaldehyde
Formula: C₁₁H₁₅NO
Boiling Pt: 174 °C/7mmHg
Melting Pt: 38…42 °C
Flash Pt: 165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003382
CAS Number: 120-21-8
UN: 3077
ADR: 9+EHS,III