5-Amino-1-methyl-1H-pyrazole 97%
Supplier: Apollo Scientific
Warning
OR8822-1G OR8822-25G OR8822-5G
APOSOR8822-1GEA 21.9 EUR
APOSOR8822-1G APOSOR8822-25G APOSOR8822-5G
5-Amino-1-methyl-1H-pyrazole 97%
5-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
Boiling Pt: 95…96 °C/0.5mmHg
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00068156
CAS Number: 1192-21-8