4-Butoxybenzonitrile 98%
Supplier: Apollo Scientific
OR900343-100G
OR900343-25G
APOSOR900343-100GEA
556
EUR
APOSOR900343-100G
APOSOR900343-25G
4-Butoxybenzonitrile 98%
4-Butoxybenzonitrile
Formula:
C₁₁H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD00043482 CAS Number: 5203-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...