4-Chromanol 97%
Supplier: Apollo Scientific
OR900753-25G OR900753-5G
APOSOR900753-25GEA 289 EUR
APOSOR900753-25G APOSOR900753-5G
4-Chromanol 97%
4-Chromanol
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00006849
CAS Number: 1481-93-2