4-Propylbenzene-1,3-diol 97%
Supplier: Apollo Scientific
Warning
OR9023-1G OR9023-250MG OR9023-5G
APOSOR9023-1GEA 69.6 EUR
APOSOR9023-1G APOSOR9023-250MG APOSOR9023-5G
4-Propylbenzene-1,3-diol 97%
4-Propylbenzene-1,3-diol
Formula: C₉H₁₂O₂
Boiling Pt: 286.9 °C
Melting Pt: 82.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00020120
CAS Number: 18979-60-7