7-Chloro-3,4-dihydroquinolin-2(1H)-one 95%
Supplier: Apollo Scientific
OR903655-1G OR903655-250MG
APOSOR903655-1GEA 139 EUR
APOSOR903655-1G APOSOR903655-250MG
7-Chloro-3,4-dihydroquinolin-2(1H)-one 95%
7-Chloro-3,4-dihydroquinolin-2(1H)-one
Formula: C₉H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD09033162
CAS Number: 14548-50-6