4-Chloro-6-methoxyquinolin-7-ol 95%

Supplier: Apollo Scientific

OR903690-1G OR903690-250MG OR903690-5G
APOSOR903690-1GEA 37.3 EUR
APOSOR903690-1G APOSOR903690-250MG APOSOR903690-5G
4-Chloro-6-methoxyquinolin-7-ol 95%
4-Chloro-6-methoxyquinolin-7-ol
Formula: C₁₀H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD13191788
CAS Number: 205448-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR