2-(4-Aminophenyl)-2-methylpropan-1-ol 95%
Supplier: Apollo Scientific
Warning
OR903940-100MG OR903940-250MG
APOSOR903940-100MGEA 230 EUR
APOSOR903940-100MG APOSOR903940-250MG
2-(4-Aminophenyl)-2-methylpropan-1-ol 95%
2-(4-Aminophenyl)-2-methylpropan-1-ol
Formula: C₁₀H₁₅NO
Boiling Pt: 129…131.5 °C
Storage Temperature: Ambient
CAS Number: 18755-56-1