Ethyl-3-bromopropionate 97%
Supplier: Apollo Scientific
Warning
OR904910-100G OR904910-500G
APOSOR904910-100GEA 24.4 EUR
APOSOR904910-100G APOSOR904910-500G
Ethyl-3-bromopropionate 97%
Ethyl 3-bromopropionate
Formula: C₅H₉BrO₂
Boiling Pt: 135…136 °C
Density: 1.412 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000251
CAS Number: 539-74-2