2-Hydroxy-4-methoxybenzonitrile 95%
Supplier: Apollo Scientific
OR907186-1G OR907186-250MG OR907186-5G
APOSOR907186-1GEA 89.9 EUR
APOSOR907186-1G APOSOR907186-250MG APOSOR907186-5G
2-Hydroxy-4-methoxybenzonitrile 95%
2-Hydroxy-4-methoxybenzonitrile
Formula: C₈H₇NO₂
MW: 149.1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08567959
CAS Number: 39835-11-5