1-Ethyl-1H-pyrazole-4-boronic acid 98%
Supplier: Apollo Scientific
Synonyms: 1-Ethylpyrazole-4-boronic Acid
OR907490-1G OR907490-5G
APOSOR907490-1GEA 183 EUR
APOSOR907490-1G APOSOR907490-5G
1-Ethyl-1H-pyrazole-4-boronic acid 98%
1-Ethyl-1H-pyrazole-4-boronic acid
Formula: C₅H₉BN₂O₂
MW: 139.9500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703521
CAS Number: 847818-56-8