N,N-Diethyl 2-methylbenzamide 98%
Supplier: Apollo Scientific
OR907818-1G OR907818-5G
APOSOR907818-1GEA 168 EUR
APOSOR907818-1G APOSOR907818-5G
N,N-Diethyl 2-methylbenzamide 98%
N,N-Diethyl 2-methylbenzamide
Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD00463019
CAS Number: 2728-04-3