3-Chloro-2-methylbenzamide 95%
Supplier: Apollo Scientific
OR908180-25G
OR908180-5G
APOSOR908180-25GEA
114
EUR
APOSOR908180-25G
APOSOR908180-5G
3-Chloro-2-methylbenzamide 95%
3-Chloro-2-methylbenzamide
Formula:
C₈H₈ClNO Storage Temperature: Ambient |
MDL Number:
MFCD00047882 CAS Number: 205178-79-6 |