tert-Butyl-3-hydroxyazetidine-1-carboxylate 97%
Supplier: Apollo Scientific
Danger
Synonyms: N-Boc-3-hydroxyazetidine, tert-Butyl 3-hydroxyazetidine-1-carboxylate, 1-BOC-3-Hydroxyazetidine, tert-butyl 3-hydroxyazetidine-1-carboxylate
OR9092-25G OR9092-100G
APOSOR9092-25GEA 71.6 EUR
APOSOR9092-25G APOSOR9092-100G
tert-Butyl-3-hydroxyazetidine-1-carboxylate 97%
tert-Butyl-3-hydroxyazetidine-1-carboxylate
Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Melting Pt: 36…47 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115305
CAS Number: 141699-55-0