1-(4-Cyanophenyl)-2,5-dimethylpyrrole 98%
Supplier: Apollo Scientific
Synonyms: 4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile
OR909537-10G OR909537-1G OR909537-25G
APOSOR909537-10GEA 342 EUR
APOSOR909537-10G APOSOR909537-1G APOSOR909537-25G
1-(4-Cyanophenyl)-2,5-dimethylpyrrole 98%
1-(4-Cyanophenyl)-2,5-dimethylpyrrole
Formula: C₁₃H₁₂N₂
MW: 196.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03228548
CAS Number: 119516-86-8