4-Methyl-5-vinylthiazole 95%

Supplier: Apollo Scientific

OR911543-100G OR911543-25G
APOSOR911543-100GEA 52.9 EUR
APOSOR911543-100G APOSOR911543-25G
4-Methyl-5-vinylthiazole 95%
4-Methyl-5-vinylthiazole
Formula: C₆H₇NS
Storage Temperature: Ambient
MDL Number: MFCD00005337
CAS Number: 1759-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR