1-Bromo-3-methoxypropane 97%
Supplier: Apollo Scientific
Warning
OR911631-100G OR911631-25G
APOSOR911631-100GEA 43.5 EUR
APOSOR911631-100G APOSOR911631-25G
1-Bromo-3-methoxypropane 97%
1-Bromo-3-methoxypropane
Formula: C₄H₉BrO
Boiling Pt: 132 °C
Density: 1.37 g/cm³ (25 °C)
Flash Pt: <60 °C
Storage Temperature: Ambient
MDL Number: MFCD02258473
CAS Number: 36865-41-5
UN: 2344
ADR: 3,III