4-Methylthiazole 95%
Supplier: Apollo Scientific
Danger
OR911882-100G OR911882-500G
APOSOR911882-100GEA 63.8 EUR
APOSOR911882-100G APOSOR911882-500G
4-Methylthiazole 95%
4-Methylthiazole
Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 133…134 °C
Density: 1.09 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00005340
CAS Number: 693-95-8
UN: 1993
ADR: 3 III