tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 97%
Supplier: Apollo Scientific
OR912133-1G OR912133-5G
APOSOR912133-1GEA 0 EUR
APOSOR912133-1G APOSOR912133-5G
tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 97%
tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate
Formula: C₁₀H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD03426270
CAS Number: 135065-69-9