4-N-Boc-aminomethylcyclohexanone 96%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Aminomethyl)cyclohexanone, N-BOC protected, tert-Butyl ((4-oxocyclohexyl)methyl)carbamate, 4-N-Boc-aminomethyl-cyclohexanone
OR913549-1G OR913549-5G
APOSOR913549-1GEA 46.4 EUR
APOSOR913549-1G APOSOR913549-5G
4-N-Boc-aminomethylcyclohexanone 96%
4-N-Boc-aminomethylcyclohexanone
Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03839885
CAS Number: 809273-70-9