4-tert-Butoxybenzonitrile 95%

Supplier: Apollo Scientific

Synonyms: 4-(tert-Butoxy)benzonitrile

OR916364-25G OR916364-5G
APOSOR916364-25GEA 231 EUR
APOSOR916364-25G APOSOR916364-5G
4-tert-Butoxybenzonitrile 95%
4-tert-Butoxybenzonitrile
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425273
CAS Number: 185259-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR