Trichloroacetic anhydride 95%
Supplier: Apollo Scientific
OR918986-250G OR918986-50G
APOSOR918986-250GEA 536 EUR
APOSOR918986-250G APOSOR918986-50G
Trichloroacetic anhydride 95%
Trichloroacetic anhydride
Formula: C₄Cl₆O₃
MW: 308.76 g/mol
Boiling Pt: 139…141 °C/60mmHg
Melting Pt: -2
Density: 1.69 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000793
CAS Number: 4124-31-6
UN: 3265
ADR: 8 I