2,5-Dichlorophenylhydrazine
Supplier: Apollo Scientific
Danger
OR9202-25G OR9202-5G
APOSOR9202-25GEA 125 EUR
APOSOR9202-25G APOSOR9202-5G
2,5-Dichlorophenylhydrazine
2,5-Dichlorophenylhydrazine
Formula: C₆H₆Cl₂N₂
Melting Pt: 102…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00007576
CAS Number: 305-15-7
UN: 2811
ADR: 6.1,III