2-Morpholin-4-ylmethylbenzylamine 95+%
Supplier: Apollo Scientific
Synonyms: 1-[2-(Morpholin-4-ylmethyl)phenyl]methylamine
OR9210-10G OR9210-1G OR9210-5G
APOSOR9210-10GEA 821 EUR
APOSOR9210-10G APOSOR9210-1G APOSOR9210-5G
2-Morpholin-4-ylmethylbenzylamine 95+%
2-Morpholin-4-ylmethylbenzylamine
Formula: C₁₂H₁₈N₂O
MW: 206.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655814
CAS Number: 91271-82-8