1,2-Diethoxyethane 95%
Supplier: Apollo Scientific
Synonyms: Diethyl glycol, Ethylene glycol diethyl ether
OR921322-100G OR921322-1KG OR921322-500G
APOSOR921322-100GEA 125 EUR
APOSOR921322-100G APOSOR921322-1KG APOSOR921322-500G
1,2-Diethoxyethane 95%
1,2-Diethoxyethane
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 121 °C/760mmHg
Melting Pt: –74 °C
Density: 0.842 g/cm³ (25 °C)
Flash Pt: 19 °C
Storage Temperature: Ambient
MDL Number: MFCD00009253
CAS Number: 629-14-1
UN: 1153
ADR: 3 II