Nitrilotriacetic acid 99%
Supplier: Apollo Scientific
Synonyms: NTA, 2,2',2''-Nitrilotriacetic acid
OR921592-2.5KG OR921592-500G
APOSOR921592-2.5KGEA 388 EUR
APOSOR921592-2.5KG APOSOR921592-500G
Nitrilotriacetic acid 99%
Nitrilotriacetic acid
Formula: C₆H₉NO₆
MW: 191.14 g/mol
Boiling Pt: 498
Melting Pt: 242
Storage Temperature: Ambient
MDL Number: MFCD00004287
CAS Number: 139-13-9
Merck Index: 12,06675