Methyl-3-(dimethylamino)benzoate 98%
Supplier: Apollo Scientific
Synonyms: 3-(Dimethylamino)benzoic acid methyl ester
OR922281-100G OR922281-25G
APOSOR922281-100GEA 831 EUR
APOSOR922281-100G APOSOR922281-25G
Methyl-3-(dimethylamino)benzoate 98%
Methyl 3-(dimethylamino)benzoate
Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00144731
CAS Number: 16518-64-2