sec-Butyl ethyl ether 95%
Supplier: Apollo Scientific
OR922803-25G OR922803-5G
APOSOR922803-25GEA 0 EUR
APOSOR922803-25G APOSOR922803-5G
sec-Butyl ethyl ether 95%
sec-Butyl ethyl ether
Formula: C₆H₁₄O
MW: 102.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053773
CAS Number: 2679-87-0