1-[3-(Dimethylamino)propyl]-3-ethylurea 98%
Supplier: Apollo Scientific
OR922815-1G OR922815-5G
APOSOR922815-1GEA 0 EUR
APOSOR922815-1G APOSOR922815-5G
1-[3-(Dimethylamino)propyl]-3-ethylurea 98%
1-[3-(Dimethylamino)propyl]-3-ethylurea
Formula: C₈H₁₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD00027752
CAS Number: 32897-26-0