3-TYP 97%
Supplier: Apollo Scientific
OR923030-1G
OR923030-250MG
APOSOR923030-1GEA
579
EUR
APOSOR923030-1G
APOSOR923030-250MG
3-TYP 97%
3-TYP
Formula:
C₇H₆N₄ Storage Temperature: Ambient |
MDL Number:
MFCD25956467 CAS Number: 120241-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...