Diethylaminoacetonitrile 95%
Supplier: Apollo Scientific
Synonyms:
N,N-Diethylglycinonitrile
OR923081-25G
OR923081-5G
APOSOR923081-25GEA
342
EUR
APOSOR923081-25G
APOSOR923081-5G
Diethylaminoacetonitrile 95%
Diethylaminoacetonitrile
Formula:
C₆H₁₂N₂ MW: 112.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00001891 CAS Number: 3010-02-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...