Barium hydroxide 94+%
Supplier: Apollo Scientific
Danger
OR923866-1KG OR923866-250G OR923866-50G
APOSOR923866-1KGEA 185 EUR
APOSOR923866-1KG APOSOR923866-250G APOSOR923866-50G
Barium hydroxide 94+%
Barium hydroxide
Formula: H₂BaO₂
MW: 171.34 g/mol
Melting Pt: 325
Density: 2.18
Storage Temperature: Ambient
MDL Number: MFCD00003443
CAS Number: 17194-00-2
UN: 3262
ADR: 8 II
Merck Index: 12,01006