Benzyl alcohol
Supplier: Apollo Scientific
Warning
APOSOR923999-1KGEA 94.5 EUR
APOSOR923999-1KG APOSOR923999-10KG
Benzyl alcohol
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108,14 g/mol
Boiling Pt: 205 °C
Melting Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6