3-Methoxy-3-methyl-1-butanol 95%

Supplier: Apollo Scientific

OR925046-100G OR925046-500G
APOSOR925046-100GEA 23 EUR
APOSOR925046-100G APOSOR925046-500G
3-Methoxy-3-methyl-1-butanol 95%
3-Methoxy-3-methyl-1-butanol
Formula: C₆H₁₄O₂
Boiling Pt: 173…175 °C
Melting Pt: –50 °C
Density: 0.926 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044771
CAS Number: 56539-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR