trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 95%
Supplier: Apollo Scientific
Synonyms: trans, trans-2,4-Hexadienedioic acid
OR925263-10G OR925263-1G OR925263-5G
APOSOR925263-10GEA 0 EUR
APOSOR925263-10G APOSOR925263-1G APOSOR925263-5G
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 95%
trans,trans-Muconic acid (trans,trans-1,3-Butadiene-1,4-dicarboxylic acid)
Formula: C₆H₆O₄
MW: 142.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002702
CAS Number: 3588-17-8