3-Cyclopentylpropionyl chloride 95%
Supplier: Apollo Scientific
OR925382-100G
OR925382-25G
APOSOR925382-100GEA
169
EUR
APOSOR925382-100G
APOSOR925382-25G
3-Cyclopentylpropionyl chloride 95%
3-Cyclopentylpropionyl chloride
Formula:
C₈H₁₃ClO Storage Temperature: Ambient |
MDL Number:
MFCD00001391 CAS Number: 104-97-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...